2-Bromo-4-(1-(4-fluorophenyl)ethyl)-1-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

48129-500MG 48129-1G 48129-5G
77337-356EA 412 USD
77337-356 77351-056 77375-396
2-Bromo-4-(1-(4-fluorophenyl)ethyl)-1-methoxybenzene ≥95%
2-Bromo-4-(1-(4-fluorophenyl)ethyl)-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrFO
MW: 309.173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707299
CAS Number: 2484888-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR