2-Bromo-1-methoxy-4-(1-(p-tolyl)ethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

48128-500MG 48128-1G 48128-5G
77336-722EA 393.46 USD
77336-722 77351-054 77375-394
2-Bromo-1-methoxy-4-(1-(p-tolyl)ethyl)benzene ≥97%
2-Bromo-1-methoxy-4-(1-(p-tolyl)ethyl)benzene

Precaución: for Research Use Only

Formula: C₁₆H₁₇BrO
MW: 305.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707298
CAS Number: 2484889-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR