4-(5-Bromo-2-fluorobenzyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48123-500MG 48123-1G 48123-5G
77327-464EA 338.87 USD
77327-464 77351-052 77375-392
4-(5-Bromo-2-fluorobenzyl)benzaldehyde ≥97%
4-(5-Bromo-2-fluorobenzyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrFO
MW: 293.131 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902729
CAS Number: 2504203-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR