Ethyl 4-bromo-2-methylsulfanylbenzoate ≥97%

Supplier: AOBChem USA

Warning

48114-500MG 48114-1G 48114-5G
77327-462EA 322.39 USD
77327-462 77351-046 77375-386
Ethyl 4-bromo-2-methylsulfanylbenzoate ≥97%
Ethyl 4-bromo-2-methylsulfanylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD22488928
CAS Number: 918328-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR