4-Bromo-2-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

48113-500MG 48113-1G 48113-5G
77327-460EA 307.97 USD
77327-460 77351-044 77375-384
4-Bromo-2-(methylthio)benzoic acid ≥97%
4-Bromo-2-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₂S
Storage Temperature: Ambient
CAS Number: 255727-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR