3-Bromo-4-fluoro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

48108-500MG 48108-1G 48108-5G 48108-25G
77327-458EA 145.23 USD
77327-458 77345-074 77363-808 77386-264
3-Bromo-4-fluoro-N-methoxy-N-methylbenzamide ≥97%
3-Bromo-4-fluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD11847401
CAS Number: 1220124-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR