(3-Bromo-4-fluorophenyl)(5-chloropyridin-2-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

48107-500MG 48107-1G 48107-5G
77327-456EA 336.81 USD
77327-456 77351-042 77375-382
(3-Bromo-4-fluorophenyl)(5-chloropyridin-2-yl)methanone ≥95%
(3-Bromo-4-fluorophenyl)(5-chloropyridin-2-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₆BrClFNO
MW: 314.538 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403873
CAS Number: 2097065-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR