3-Bromo-5-fluoro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

48106-500MG 48106-1G 48106-5G 48106-25G
77327-454EA 231.75 USD
77327-454 77351-040 77363-806 77386-262
3-Bromo-5-fluoro-N-methoxy-N-methylbenzamide ≥97%
3-Bromo-5-fluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD18840628
CAS Number: 1339239-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR