(5-(methylthio)pyridin-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

48093-500MG 48093-1G 48093-5G
77338-714EA 511.91 USD
77338-714 77351-034 77375-376
(5-(methylthio)pyridin-3-yl)methanol ≥97%
(5-(methylthio)pyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₉NOS
Storage Temperature: Ambient
MDL Number: MFCD28680923
CAS Number: 1173713-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR