1-Bromo-3-methoxy-5-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

48090-500MG 48090-1G 48090-5G 48090-25G
77327-452EA 176.13 USD
77327-452 77346-524 77363-804 77386-260
1-Bromo-3-methoxy-5-(methoxymethyl)benzene ≥97%
1-Bromo-3-methoxy-5-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD34529966
CAS Number: 2734772-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR