(5-Bromo-2,3-difluoro-4-methylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

48085-500MG 48085-1G 48085-5G 48085-25G
77327-450EA 223.51 USD
77327-450 77351-032 77363-802 77386-258
(5-Bromo-2,3-difluoro-4-methylphenyl)methanol ≥95%
(5-Bromo-2,3-difluoro-4-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O
MW: 237.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707294
CAS Number: 2484889-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR