5-Bromo-2-fluoro-3-methoxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

48079-500MG 48079-1G 48079-5G
77336-614EA 390.37 USD
77336-614 77351-030 77375-372
5-Bromo-2-fluoro-3-methoxybenzonitrile ≥97%
5-Bromo-2-fluoro-3-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFNO
MW: 230.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902726
CAS Number: 1898131-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR