(3-Bromo-2,6-dichlorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

48078-500MG 48078-1G 48078-5G
77335-500EA 356.38 USD
77335-500 77351-028 77375-370
(3-Bromo-2,6-dichlorophenyl)methanol ≥97%
(3-Bromo-2,6-dichlorophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD20923582
CAS Number: 1807171-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR