(2,3-difluoro-6-(methylthio)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

48065-500MG 48065-1G 48065-25G 48065-5G
77327-442EA 296.64 USD
77327-442 77351-020 77386-250 77375-362
(2,3-difluoro-6-(methylthio)phenyl)methanol ≥97%
(2,3-difluoro-6-(methylthio)phenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈F₂OS
Storage Temperature: Ambient
MDL Number: MFCD34564110
CAS Number: 2734772-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR