6-Chloro-4-methylpicolinaldehyde ≥97%

Supplier: AOBChem USA

Warning

48061-500MG 48061-1G 48061-5G
77338-948EA 534.57 USD
77338-948 77351-016 77375-360
6-Chloro-4-methylpicolinaldehyde ≥97%
6-Chloro-4-methylpicolinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆ClNO
MW: 155.582 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189134
CAS Number: 1060804-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR