Ethyl 4-bromo-2,3-dichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

48058-500MG 48058-1G 48058-5G 48058-25G
77327-438EA 123.6 USD
77327-438 77344-034 77363-790 77375-358
Ethyl 4-bromo-2,3-dichlorobenzoate ≥97%
Ethyl 4-bromo-2,3-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrCl₂O₂
MW: 297.961 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178538
CAS Number: 2386342-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR