5-Bromo-3-ethoxy-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48051-500MG 48051-1G 48051-5G 48051-25G
77327-434EA 177.16 USD
77327-434 77346-590 77363-786 77386-244
5-Bromo-3-ethoxy-2-fluorobenzaldehyde ≥97%
5-Bromo-3-ethoxy-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902719
CAS Number: 2386234-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR