2-Fluoro-3-methoxy-4-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

48048-500MG 48048-1G 48048-5G 48048-25G
77327-428EA 106.09 USD
77327-428 77343-126 77363-782 77375-350
2-Fluoro-3-methoxy-4-(methylthio)pyridine ≥97%
2-Fluoro-3-methoxy-4-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₈FNOS
MW: 173.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403871
CAS Number: 2624417-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR