2,3-dimethoxy-4-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

48043-500MG 48043-1G 48043-5G 48043-25G
77327-426EA 179.22 USD
77327-426 77346-714 77363-780 77386-240
2,3-dimethoxy-4-(methylthio)pyridine ≥97%
2,3-dimethoxy-4-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂S
MW: 185.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564108
CAS Number: 2734774-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR