(2,3-dimethoxypyridin-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

48041-500MG 48041-1G 48041-5G
77338-644EA 506.76 USD
77338-644 77351-002 77375-346
(2,3-dimethoxypyridin-4-yl)methanol ≥97%
(2,3-dimethoxypyridin-4-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₁₁NO₃
MW: 169.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28518794
CAS Number: 1783326-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR