3-Bromo-2,6-difluoro-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

47994-500MG 47994-1G 47994-5G
77337-650EA 434.66 USD
77337-650 77350-996 77375-338
3-Bromo-2,6-difluoro-5-methoxybenzaldehyde ≥97%
3-Bromo-2,6-difluoro-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34178534
CAS Number: 1785601-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR