3-Bromo-2,6-difluoro-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

47992-500MG 47992-1G 47992-5G
77337-350EA 412 USD
77337-350 77350-994 77375-336
3-Bromo-2,6-difluoro-5-methoxybenzoic acid ≥97%
3-Bromo-2,6-difluoro-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₃
MW: 267.024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178533
CAS Number: 2643368-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR