Ethyl 2-chloro-3,4-difluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

47972-500MG 47972-1G 47972-5G 47972-25G
77327-424EA 280.16 USD
77327-424 77350-990 77363-776 77386-238
Ethyl 2-chloro-3,4-difluorobenzoate ≥95%
Ethyl 2-chloro-3,4-difluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇ClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18416422
CAS Number: 1261541-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR