Ethyl 3,5-difluoro-4-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

47971-500MG 47971-1G 47971-5G 47971-25G
77327-422EA 150.38 USD
77327-422 77345-340 77363-774 77386-236
Ethyl 3,5-difluoro-4-methylbenzoate ≥95%
Ethyl 3,5-difluoro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₂
MW: 200.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28790312
CAS Number: 1806321-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR