1,3-Difluoro-4-iodo-2-methylsulfanylbenzene ≥97%

Supplier: AOBChem USA

Warning

47957-1G 47957-5G 47957-25G
77341-758EA 104.03 USD
77341-758 77363-768 77375-330
1,3-Difluoro-4-iodo-2-methylsulfanylbenzene ≥97%
1,3-Difluoro-4-iodo-2-methylsulfanylbenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂IS
Storage Temperature: Ambient
MDL Number: MFCD32707289
CAS Number: 2484889-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR