Methyl 3,5-dichloro-4-propoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

47951-500MG 47951-1G 47951-5G 47951-25G
77327-414EA 114.33 USD
77327-414 77343-358 77363-766 77375-328
Methyl 3,5-dichloro-4-propoxybenzoate ≥97%
Methyl 3,5-dichloro-4-propoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD31774417
CAS Number: 1368285-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR