Ethyl 3,5-dichloro-4-ethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

47948-500MG 47948-1G 47948-5G
77337-348EA 412 USD
77337-348 77350-982 77375-324
Ethyl 3,5-dichloro-4-ethoxybenzoate ≥97%
Ethyl 3,5-dichloro-4-ethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂Cl₂O₃
MW: 263.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07021304
CAS Number: 40689-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR