(3,5-Dichloro-4-ethoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

47947-500MG 47947-1G 47947-5G
77337-786EA 443.93 USD
77337-786 77350-980 77375-322
(3,5-Dichloro-4-ethoxyphenyl)methanol ≥97%
(3,5-Dichloro-4-ethoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03862359
CAS Number: 664319-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR