4-(thiophen-2-ylmethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

47928-500MG 47928-1G 47928-5G
77327-410EA 338.87 USD
77327-410 77350-978 77375-320
4-(thiophen-2-ylmethyl)benzaldehyde ≥97%
4-(thiophen-2-ylmethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₀OS
MW: 202.272 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13151980
CAS Number: 1007847-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR