tert-Butyl 4-(p-tolyl)piperidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

47918-500MG 47918-1G 47918-5G
77337-266EA 410.97 USD
77337-266 77350-974 77375-318
tert-Butyl 4-(p-tolyl)piperidine-1-carboxylate ≥95%
tert-Butyl 4-(p-tolyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD11111400
CAS Number: 732275-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR