4-Methyl-5-phenylpyridin-2-amine ≥97%

Supplier: AOBChem USA

Warning

47872-500MG 47872-1G 47872-5G
77337-576EA 424.36 USD
77337-576 77350-972 77375-316
4-Methyl-5-phenylpyridin-2-amine ≥97%
4-Methyl-5-phenylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂
Storage Temperature: Ambient
CAS Number: 84596-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR