1,5-Dichloro-2-iodo-3-nitrobenzene ≥95%

Supplier: AOBChem USA

Warning

47775-500MG 47775-1G 47775-5G 47775-25G
77327-404EA 189.52 USD
77327-404 77350-970 77363-758 77386-226
1,5-Dichloro-2-iodo-3-nitrobenzene ≥95%
1,5-Dichloro-2-iodo-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₂INO₂
Storage Temperature: Ambient
MDL Number: MFCD08275978
CAS Number: 189576-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR