N-Cyclopropyl-3,5-bis(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

47718-1G 47718-5G 47718-25G
77341-620EA 96.82 USD
77341-620 77363-756 77375-314
N-Cyclopropyl-3,5-bis(trifluoromethyl)benzamide ≥97%
N-Cyclopropyl-3,5-bis(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₂H₉F₆NO
Storage Temperature: Ambient
MDL Number: MFCD26687303
CAS Number: 1244040-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR