N-Cyclohexyl-3,5-bis(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

47717-1G 47717-5G 47717-25G
77341-618EA 96.82 USD
77341-618 77363-754 77375-312
N-Cyclohexyl-3,5-bis(trifluoromethyl)benzamide ≥97%
N-Cyclohexyl-3,5-bis(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₅H₁₅F₆NO
MW: 339.276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01489479
CAS Number: 1244040-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR