2-(3-Ethoxy-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%

Supplier: AOBChem USA

Warning

47704-500MG 47704-1G 47704-5G
77327-400EA 332.69 USD
77327-400 77350-968 77375-306
2-(3-Ethoxy-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%
2-(3-Ethoxy-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₅H₂₃BO₃
Storage Temperature: Ambient
MDL Number: MFCD32902712
CAS Number: 2246830-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR