4-Methyl-N'-((1-Methyl-1H-pyrazol-4-yl)methylene)benzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47686-1G 47686-5G 47686-25G
77341-240EA 78.28 USD
77341-240 77363-748 77375-302
4-Methyl-N'-((1-Methyl-1H-pyrazol-4-yl)methylene)benzenesulfonohydrazide ≥97%
4-Methyl-N'-((1-Methyl-1H-pyrazol-4-yl)methylene)benzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₄O₂S
MW: 278.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707284
CAS Number: 1287383-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR