N'-(1-(2,5-difluorophenyl)ethylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47684-1G 47684-5G 47684-25G
77341-014EA 66.95 USD
77341-014 77363-746 77375-300
N'-(1-(2,5-difluorophenyl)ethylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(1-(2,5-difluorophenyl)ethylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₅H₁₄F₂N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD11264881
CAS Number: 1931949-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR