N'-(6-Bromo-2,3-dihydro-1H-inden-1-ylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47682-1G 47682-5G 47682-25G
77341-012EA 66.95 USD
77341-012 77363-744 77375-298
N'-(6-Bromo-2,3-dihydro-1H-inden-1-ylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(6-Bromo-2,3-dihydro-1H-inden-1-ylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₆H₁₅BrN₂O₂S
MW: 379.272 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707282
CAS Number: 1978295-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR