2-(3,5-bis(trifluoromethyl)benzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

47638-500MG 47638-1G 47638-5G 47638-25G
77327-396EA 126.69 USD
77327-396 77344-112 77363-740 77375-296
2-(3,5-bis(trifluoromethyl)benzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(3,5-bis(trifluoromethyl)benzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₅H₁₇BF₆O₂
MW: 354.096 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756383
CAS Number: 1854115-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR