5-(Benzyloxy)-2,4-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

47542-500MG 47542-1G 47542-5G
77336-672EA 391.4 USD
77336-672 77350-958 77375-288
5-(Benzyloxy)-2,4-difluorobenzaldehyde ≥97%
5-(Benzyloxy)-2,4-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34178530
CAS Number: 2643367-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR