2-(4-Bromo-2,6-difluorophenyl)-2-methyl-1,3-dioxolane ≥97%
Supplier: AOBChem USA
|
Warning
|
47540-500MG
47540-1G
47540-5G
47540-25G
77327-392EA
101.97
USD
77327-392
77342-994
77363-738
77375-286
2-(4-Bromo-2,6-difluorophenyl)-2-methyl-1,3-dioxolane ≥97%
2-(4-Bromo-2,6-difluorophenyl)-2-methyl-1,3-dioxolane
Precaución: for Research Use Only
Formula:
C₁₀H₉BrF₂O₂ MW: 279.078 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32707280 CAS Number: 2484889-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...