Ethyl 2,6-difluoro-3-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

47521-500MG 47521-1G 47521-5G 47521-25G
77327-386EA 147.29 USD
77327-386 77345-198 77363-732 77386-218
Ethyl 2,6-difluoro-3-isopropoxybenzoate ≥97%
Ethyl 2,6-difluoro-3-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34564105
CAS Number: 2734774-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR