(4-(Benzyloxy)-3-methylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

47514-500MG 47514-1G 47514-5G 47514-25G
77327-384EA 243.08 USD
77327-384 77350-948 77363-728 77386-216
(4-(Benzyloxy)-3-methylphenyl)methanol ≥95%
(4-(Benzyloxy)-3-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₂
MW: 228.286 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403859
CAS Number: 1898255-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR