Methyl 4-(benzyloxy)-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

47512-500MG 47512-1G 47512-5G 47512-25G
77327-380EA 259.56 USD
77327-380 77350-944 77363-724 77386-212
Methyl 4-(benzyloxy)-3-methylbenzoate ≥97%
Methyl 4-(benzyloxy)-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
MW: 256.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26401600
CAS Number: 1439364-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR