1-Bromo-3-chloro-4-ethoxy-2-(methoxymethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

47510-500MG 47510-1G 47510-5G 47510-25G
77327-378EA 191.58 USD
77327-378 77350-942 77363-722 77386-210
1-Bromo-3-chloro-4-ethoxy-2-(methoxymethyl)benzene ≥95%
1-Bromo-3-chloro-4-ethoxy-2-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrClO₂
MW: 279.558 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707278
CAS Number: 2484888-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR