(6-Bromo-2-chloro-3-ethoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

47509-500MG 47509-1G 47509-5G 47509-25G
77327-376EA 150.38 USD
77327-376 77345-338 77363-720 77386-208
(6-Bromo-2-chloro-3-ethoxyphenyl)methanol ≥95%
(6-Bromo-2-chloro-3-ethoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO₂
MW: 265.531 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707277
CAS Number: 2386676-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR