2-(6-Bromo-2-chloro-3-ethoxyphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

47507-500MG 47507-1G 47507-5G 47507-25G
77327-372EA 136.99 USD
77327-372 77344-528 77363-718 77386-204
2-(6-Bromo-2-chloro-3-ethoxyphenyl)-1,3-dioxolane ≥95%
2-(6-Bromo-2-chloro-3-ethoxyphenyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrClO₃
Storage Temperature: Ambient
MDL Number: MFCD32707276
CAS Number: 2484889-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR