Ethyl 6-bromo-2-chloro-3-ethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

47506-500MG 47506-1G 47506-5G 47506-25G
77327-370EA 126.69 USD
77327-370 77344-110 77363-716 77375-276
Ethyl 6-bromo-2-chloro-3-ethoxybenzoate ≥97%
Ethyl 6-bromo-2-chloro-3-ethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrClO₃
MW: 307.568 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32874088
CAS Number: 2384335-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR