4-(Benzyloxy)-3-ethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

47444-500MG 47444-1G 47444-5G 47444-25G
77327-356EA 225.57 USD
77327-356 77350-932 77363-698 77386-198
4-(Benzyloxy)-3-ethylbenzaldehyde ≥97%
4-(Benzyloxy)-3-ethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₂
MW: 240.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403854
CAS Number: 886230-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR