Methyl 4-(benzyloxy)-3-ethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

47443-500MG 47443-1G 47443-5G
77327-354EA 320.33 USD
77327-354 77350-930 77375-260
Methyl 4-(benzyloxy)-3-ethylbenzoate ≥97%
Methyl 4-(benzyloxy)-3-ethylbenzoate

Precaución: for Research Use Only

Formula: C₁₇H₁₈O₃
MW: 270.323 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756372
CAS Number: 2901082-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR